Assay Design



Gene Transcripts

Gene Symbol
CCL28
Entrez Gene ID
102176270
Gene Name
C-C motif chemokine ligand 28
Gene Aliases
-
Chromosome Location
Chr.20: 31349801 - 31365879 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694738.3 XP_005694795.1 1 - 2 148 75
GenBank mRNA JO443631.2 - 1 - 2 221 75

View Full Product Details