Assay Design



Gene Transcripts

Gene Symbol
MARCH11
Entrez Gene ID
102176271
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
-
Chromosome Location
Chr.20: 56903698 - 57019252 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065811.1 XP_017921300.1 2 - 3 904 84

View Full Product Details