Assay Design



Gene Transcripts

Gene Symbol
TBRG4
Entrez Gene ID
102181265
Gene Name
transforming growth factor beta regulator 4
Gene Aliases
-
Chromosome Location
Chr.4: 43648243 - 43658631 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679316.3 XP_005679373.1 9 - 10 1826 86
XM_013963385.2 XP_013818839.1 9 - 10 1743 86

View Full Product Details