Assay Design



Gene Transcripts

Gene Symbol
MLANA
Entrez Gene ID
102178069
Gene Name
melan-A
Gene Aliases
-
Chromosome Location
Chr.8: 38632728 - 38645055 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683687.2 XP_005683744.1 2 - 3 152 64
GenBank mRNA KA320474.1 - 2 - 3 166 64

View Full Product Details