Assay Design



Gene Transcripts

Gene Symbol
LHFPL5
Entrez Gene ID
102177481
Gene Name
LHFPL tetraspan subfamily member 5
Gene Aliases
-
Chromosome Location
Chr.23: 38921502 - 38936368 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973773.2 XP_013829227.1 1 - 2 642 58
GenBank mRNA JO627591.2 - 3 - 4 405 58

View Full Product Details