Assay Design



Gene Transcripts

Gene Symbol
PGBD1
Entrez Gene ID
102171287
Gene Name
piggyBac transposable element derived 1
Gene Aliases
-
Chromosome Location
Chr.23: 19518924 - 19540136 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013974051.2 XP_013829505.2 5 - 6 3471 105
XM_013974052.2 XP_013829506.2 6 - 7 1780 105
XM_013974054.2 XP_013829508.2 6 - 7 1594 105

View Full Product Details