Assay Design



Gene Transcripts

Gene Symbol
C18H19orf54
Entrez Gene ID
102189787
Gene Name
chromosome 18 C19orf54 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 50906189 - 50914422 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692419.3 XP_005692476.1 3 - 4 1207 66

View Full Product Details