Assay Design



Gene Transcripts

Gene Symbol
LTB
Entrez Gene ID
102186643
Gene Name
lymphotoxin beta
Gene Aliases
-
Chromosome Location
Chr.23: 22250704 - 22253633 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696604.3 XP_005696661.1 2 - 3 1122 117

View Full Product Details