Assay Design



Gene Transcripts

Gene Symbol
SLC16A9
Entrez Gene ID
102188921
Gene Name
solute carrier family 16 member 9
Gene Aliases
-
Chromosome Location
Chr.28: 30120135 - 30200295 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042601.1 XP_017898090.1 2 - 3 336 79

View Full Product Details