Assay Design



Gene Transcripts

Gene Symbol
IBA57
Entrez Gene ID
102183229
Gene Name
iron-sulfur cluster assembly factor IBA57
Gene Aliases
-
Chromosome Location
Chr.7: 105853631 - 105864196 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051578.1 XP_017907067.1 2 - 3 783 103
GenBank mRNA KA314592.1 - 2 - 3 539 103

View Full Product Details