Assay Design



Gene Transcripts

Gene Symbol
LOC102190399
Entrez Gene ID
102190399
Gene Name
podocan-like protein 1
Gene Aliases
PODNL1
Chromosome Location
Chr.7: 97148147 - 97152921 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051210.1 XP_017906699.1 2 - 3 421 67
GenBank mRNA JT634544.1 - 3 - 4 517 67

View Full Product Details