Assay Design



Gene Transcripts

Gene Symbol
LOC108636402
Entrez Gene ID
108636402
Gene Name
podocan-like protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 97147970 - 97153665 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051211.1 XP_017906700.1 2 - 3 287 81

View Full Product Details