Assay Design



Gene Transcripts

Gene Symbol
TNFRSF13B
Entrez Gene ID
102184942
Gene Name
TNF receptor superfamily member 13B
Gene Aliases
-
Chromosome Location
Chr.19: 34749522 - 34772451 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971985.2 XP_013827439.2 2 - 3 415 100
XM_018064822.1 XP_017920311.1 2 - 3 415 100
GenBank mRNA JO612465.2 - 2 - 3 308 100

View Full Product Details