Assay Design



Gene Transcripts

Gene Symbol
ENSA
Entrez Gene ID
102185326
Gene Name
endosulfine alpha
Gene Aliases
-
Chromosome Location
Chr.3: 100064152 - 100071002 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677662.3 XP_005677719.1 2 - 3 331 76
GenBank mRNA JO541141.2 - - 63 76

View Full Product Details