Assay Design



Gene Transcripts

Gene Symbol
TAF5L
Entrez Gene ID
102183570
Gene Name
TATA-box binding protein associated factor 5 like
Gene Aliases
-
Chromosome Location
Chr.28: 43601312 - 43633541 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042524.1 XP_017898013.1 4 - 5 1156 62
XM_018042525.1 XP_017898014.1 5 - 6 1310 62
GenBank mRNA KA324169.1 - 4 - 5 1033 62

View Full Product Details