Assay Design



Gene Transcripts

Gene Symbol
NAALADL2
Entrez Gene ID
102170637
Gene Name
N-acetylated alpha-linked acidic dipeptidase like 2
Gene Aliases
-
Chromosome Location
Chr.1: 90886861 - 92518064 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675296.3 XP_005675353.2 8 - 9 2013 97
XM_013964525.2 XP_013819979.2 10 - 11 1767 97
GenBank mRNA KA311265.1 - 2 - 3 246 97

View Full Product Details