Assay Design



Gene Transcripts

Gene Symbol
BCL2L15
Entrez Gene ID
102169150
Gene Name
BCL2 like 15
Gene Aliases
-
Chromosome Location
Chr.3: 91090625 - 91097323 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045934.1 XP_017901423.1 2 - 3 511 75
GenBank mRNA JO332354.2 - 2 - 3 436 75

View Full Product Details