Assay Design



Gene Transcripts

Gene Symbol
TUBA8
Entrez Gene ID
102176404
Gene Name
tubulin alpha 8
Gene Aliases
-
Chromosome Location
Chr.5: 108297876 - 108316632 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048726.1 XP_017904215.1 2 - 3 411 58
GenBank mRNA JO624098.2 - 2 - 3 413 58

View Full Product Details