Assay Design



Gene Transcripts

Gene Symbol
PIGX
Entrez Gene ID
102176077
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Gene Aliases
-
Chromosome Location
Chr.1: 70957570 - 70985212 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963551.2 XP_013819005.2 3 - 4 577 109
GenBank mRNA JO438716.2 - 2 - 3 370 109

View Full Product Details