Assay Design



Gene Transcripts

Gene Symbol
ZFHX2
Entrez Gene ID
102181602
Gene Name
zinc finger homeobox 2
Gene Aliases
-
Chromosome Location
Chr.10: 80016830 - 80045550 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054606.1 XP_017910095.1 4 - 5 3174 109

View Full Product Details