Assay Design



Gene Transcripts

Gene Symbol
C6H4orf17
Entrez Gene ID
102180579
Gene Name
chromosome 6 C4orf17 homolog
Gene Aliases
-
Chromosome Location
Chr.6: 25349157 - 25443306 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049278.1 XP_017904767.1 9 - 10 1079 84
XM_018049279.1 XP_017904768.1 8 - 9 1044 84
XM_018049280.1 XP_017904769.1 8 - 9 1002 84
XM_018049281.1 XP_017904770.1 8 - 9 749 84
XM_018049282.1 XP_017904771.1 8 - 9 879 84

View Full Product Details