Assay Design



Gene Transcripts

Gene Symbol
PPP3R2
Entrez Gene ID
102177339
Gene Name
protein phosphatase 3 regulatory subunit B, beta
Gene Aliases
-
Chromosome Location
Chr.8: 91161054 - 91161947 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684257.2 XP_005684314.2 1 - 1 224 67

View Full Product Details