Assay Design



Gene Transcripts

Gene Symbol
C29H11orf86
Entrez Gene ID
102181476
Gene Name
chromosome 29 C11orf86 homolog
Gene Aliases
-
Chromosome Location
Chr.29: 45366066 - 45367567 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699942.3 XP_005699999.2 1 - 2 395 82

View Full Product Details