Assay Design



Gene Transcripts

Gene Symbol
LOC108634948
Entrez Gene ID
108634948
Gene Name
protein Dok-7-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017194558: 3063 - 13834 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045266.1 XP_017900755.1 - 334 105

Gene Symbol
DOK7
Entrez Gene ID
102182116
Gene Name
docking protein 7
Gene Aliases
-
Chromosome Location
Chr.6: 115701125 - 115712910 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049929.1 XP_017905418.1 3 - 4 440 105
GenBank mRNA JO646841.2 - - 787 105

View Full Product Details