Assay Design



Gene Transcripts

Gene Symbol
HOXB2
Entrez Gene ID
102169903
Gene Name
homeobox B2
Gene Aliases
-
Chromosome Location
Chr.19: 37579937 - 37582881 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064937.1 XP_017920426.1 1 - 2 1075 115

View Full Product Details