Assay Design



Gene Transcripts

Gene Symbol
HTR7
Entrez Gene ID
102181834
Gene Name
5-hydroxytryptamine receptor 7
Gene Aliases
-
Chromosome Location
Chr.26: 38872545 - 38969952 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041723.1 XP_017897212.1 2 - 3 1543 112

View Full Product Details