Assay Design



Gene Transcripts

Gene Symbol
AQP10
Entrez Gene ID
102180360
Gene Name
aquaporin 10
Gene Aliases
-
Chromosome Location
Chr.3: 103902046 - 103904660 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677552.2 XP_005677609.2 2 - 3 273 112

View Full Product Details