Assay Design



Gene Transcripts

Gene Symbol
VTCN1
Entrez Gene ID
102179253
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.3: 94469698 - 94544984 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677804.3 XP_005677861.2 3 - 4 513 116
XM_013962499.2 XP_013817953.2 3 - 4 513 116
GenBank mRNA KA328838.1 - 3 - 4 548 116

View Full Product Details