Assay Design



Gene Transcripts

Gene Symbol
NMBR
Entrez Gene ID
102176884
Gene Name
neuromedin B receptor
Gene Aliases
-
Chromosome Location
Chr.9: 66613446 - 66622174 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684853.3 XP_005684910.2 1 - 2 650 86

View Full Product Details