Assay Design



Gene Transcripts

Gene Symbol
TPRN
Entrez Gene ID
102178084
Gene Name
taperin
Gene Aliases
-
Chromosome Location
Chr.11: 106052822 - 106060194 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687357.3 XP_005687414.3 3 - 4 2278 113
GenBank mRNA KA343391.1 - 3 - 4 1362 113

View Full Product Details