Assay Design



Gene Transcripts

Gene Symbol
ZSCAN29
Entrez Gene ID
102180633
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
-
Chromosome Location
Chr.21: 54617439 - 54628212 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695293.3 XP_005695350.2 4 - 5 1701 75
XM_018065956.1 XP_017921445.1 3 - 4 1356 75

View Full Product Details