Assay Design



Gene Transcripts

Gene Symbol
SH2D4B
Entrez Gene ID
102179071
Gene Name
SH2 domain containing 4B
Gene Aliases
-
Chromosome Location
Chr.28: 9846200 - 9922323 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699280.3 XP_005699337.1 2 - 3 361 56

View Full Product Details