Assay Design



Gene Transcripts

Gene Symbol
NCCRP1
Entrez Gene ID
102182338
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
-
Chromosome Location
Chr.18: 49800972 - 49804829 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062383.1 XP_017917872.1 3 - 4 474 62
GenBank mRNA KA322420.1 - 3 - 4 1275 62

View Full Product Details