Assay Design



Gene Transcripts

Gene Symbol
SPINK9
Entrez Gene ID
102177234
Gene Name
serine peptidase inhibitor, Kazal type 9
Gene Aliases
-
Chromosome Location
Chr.7: 50670480 - 50677500 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683133.3 XP_005683190.2 1 - 2 157 61

View Full Product Details