Assay Design



Gene Transcripts

Gene Symbol
SLC2A4RG
Entrez Gene ID
102180608
Gene Name
SLC2A4 regulator
Gene Aliases
-
Chromosome Location
Chr.13: 53352355 - 53355917 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057500.1 XP_017912989.1 6 - 7 1166 62
GenBank mRNA JO512098.2 - 1 - 2 207 62

View Full Product Details