Assay Design



Gene Transcripts

Gene Symbol
HYAL3
Entrez Gene ID
102172770
Gene Name
hyaluronidase 3
Gene Aliases
-
Chromosome Location
Chr.22: 50114243 - 50119913 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695929.3 XP_005695986.1 2 - 3 244 80
XM_018066819.1 XP_017922308.1 2 - 3 974 80
GenBank mRNA JO588519.2 - 3 - 4 268 80

View Full Product Details