Assay Design



Gene Transcripts

Gene Symbol
SH3RF3
Entrez Gene ID
102178914
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
-
Chromosome Location
Chr.11: 43939878 - 44099310 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055296.1 XP_017910785.1 5 - 6 2294 106
XM_018055297.1 XP_017910786.1 5 - 6 2294 106

View Full Product Details