Assay Design



Gene Transcripts

Gene Symbol
BAHD1
Entrez Gene ID
102171076
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 66996638 - 67019894 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054447.1 XP_017909936.1 5 - 6 2229 65
GenBank mRNA JO558412.2 - 3 - 4 284 65

View Full Product Details