Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018057646.1 | XP_017913135.1 | 5 - 6 | 1060 | 75 |
XM_018057647.1 | XP_017913136.1 | 4 - 5 | 989 | 75 | |
XM_018057649.1 | XP_017913138.1 | 4 - 5 | 435 | 75 | |
XM_018057650.1 | XP_017913139.1 | 5 - 6 | 501 | 75 | |
GenBank mRNA | JO645466.2 | - | 4 - 5 | 465 | 75 |