Assay Design



Gene Transcripts

Gene Symbol
OSBPL10
Entrez Gene ID
102170187
Gene Name
oxysterol binding protein like 10
Gene Aliases
-
Chromosome Location
Chr.22: 6075602 - 6398911 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038204.1 XP_017893693.1 2 - 3 610 90
GenBank mRNA JO427547.1 - 10 - 11 2428 90

View Full Product Details