Assay Design



Gene Transcripts

Gene Symbol
NDUFB9
Entrez Gene ID
102188420
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
Gene Aliases
-
Chromosome Location
Chr.14: 66463013 - 66470930 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058556.1 XP_017914045.1 3 - 4 467 104
XM_018058557.1 XP_017914046.1 3 - 4 540 104
GenBank mRNA JO377275.2 - 3 - 4 461 104

View Full Product Details