Assay Design



Gene Transcripts

Gene Symbol
FAM47E
Entrez Gene ID
102184328
Gene Name
family with sequence similarity 47 member E
Gene Aliases
-
Chromosome Location
Chr.6: 91662646 - 91691932 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964737.2 XP_013820191.2 3 - 4 721 111

View Full Product Details