Assay Design



Gene Transcripts

Gene Symbol
FAM71E2
Entrez Gene ID
102184653
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
-
Chromosome Location
Chr.18: 63882140 - 63888338 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063236.1 XP_017918725.1 5 - 6 649 79

View Full Product Details