Assay Design



Gene Transcripts

Gene Symbol
MANEAL
Entrez Gene ID
102173525
Gene Name
mannosidase endo-alpha like
Gene Aliases
-
Chromosome Location
Chr.3: 12685595 - 12694129 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678678.3 XP_005678735.3 1 - 2 631 60
GenBank mRNA JO638869.2 - 3 - 4 1927 60

View Full Product Details