Assay Design



Gene Transcripts

Gene Symbol
TRAM1L1
Entrez Gene ID
102173539
Gene Name
translocation associated membrane protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.6: 8657391 - 8659570 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681281.3 XP_005681338.1 1 - 1 1253 84

View Full Product Details