Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_013969330.2 | XP_013824784.1 | 3 - 4 | 337 | 121 |
XM_013969332.2 | XP_013824786.1 | 2 - 3 | 182 | 121 | |
XM_018058207.1 | XP_017913696.1 | 3 - 4 | 351 | 121 | |
XM_018058208.1 | XP_017913697.1 | 3 - 4 | 348 | 121 | |
GenBank mRNA | JT697498.1 | - | 1 - 2 | 57 | 121 |