Assay Design



Gene Transcripts

Gene Symbol
MYMK
Entrez Gene ID
102175414
Gene Name
myomaker, myoblast fusion factor
Gene Aliases
TMEM8C
Chromosome Location
Chr.11: 103799838 - 103811935 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056155.1 XP_017911644.1 2 - 3 694 59
XM_018056156.1 XP_017911645.1 2 - 3 694 59

View Full Product Details