Assay Design



Gene Transcripts

Gene Symbol
PPP1R14D
Entrez Gene ID
102179677
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
Gene Aliases
-
Chromosome Location
Chr.10: 66693935 - 66715143 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054436.1 XP_017909925.1 4 - 5 549 103
XM_018054437.1 XP_017909926.1 3 - 4 406 103
GenBank mRNA JT711150.1 - 4 - 5 825 103

View Full Product Details