Assay Design



Gene Transcripts

Gene Symbol
C11H2orf50
Entrez Gene ID
102182137
Gene Name
chromosome 11 C2orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 86241127 - 86246560 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686999.3 XP_005687056.2 1 - 2 465 102
GenBank mRNA JO589774.2 - 2 - 3 687 102

View Full Product Details