Assay Design



Gene Transcripts

Gene Symbol
IGLON5
Entrez Gene ID
102172753
Gene Name
IgLON family member 5
Gene Aliases
-
Chromosome Location
Chr.18: 58395387 - 58436360 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063200.1 XP_017918689.1 8 - 9 1362 63
GenBank mRNA JO622870.2 - 3 - 4 443 63

View Full Product Details